Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC47A2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA524659

Catalog No. PIPA524659


Only null left
Add to Cart

Description

Description

SLC47A2 Polyclonal Antibody for Western Blot

S47A2 belongs to a family of transporters involved in excretion of toxic electrolytes, both endogenous and exogenous, through urine and bile. This transporter family shares homology with the bacterial MATE (multidrug and toxin extrusion) protein family responsible for drug resistance. This gene is one of two members of the MATE transporter family located near each other on chromosome 17.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SLC47A2
Polyclonal
Unconjugated
Slc47a2
4933429E10Rik; FLJ31196; H(+)/organic cation antiporter kidney-specific; hMATE-2; kidney-specific H(+)/organic cation antiporter; Mate2; MATE-2; MATE2-B; MATE2K; MATE2-K; mMATE-2; multidrug and toxin extrusion protein 2; SLC47A2; solute carrier family 47 (multidrug and toxin extrusion), member 2; solute carrier family 47 member 2; solute carrier family 47, member 2
Rabbit
Antigen Affinity Chromatography, Protein A
RUO
146802, 497921
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q86VL8
Slc47a2
KLH conjugated synthetic peptide between 125-153 amino acids from the N-terminal region of human S47A2.
400 μL
Primary
Human, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.