Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC48A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542191
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542191 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542191 Supplier Invitrogen™ Supplier No. PA542191
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LISGDSPASA FQSAGIIGVS HRARPGSVFL ARSEESLYLR PGQQSQEVKV.

Heme transporter that regulates intracellular heme availability through the endosomal or lysosomal compartment. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC48A1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC48A1
Gene Accession No. Q6P1K1
Gene Alias heme transporter HRG1; Heme-responsive gene 1 protein homolog; hHRG-1; HRG1; HRG-1; SLC48A1; solute carrier family 48 (heme transporter), member 1; solute carrier family 48 member 1
Gene Symbols SLC48A1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human FLJ20489.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55652
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.