Learn More
Invitrogen™ SLC4A1 Polyclonal Antibody

Description
Immunogen sequence: LLKHSHAGEL EALGGVKPAV LTRSGDPSQP LLPQHSSLET QLFCEQGDGG TEGHSPSGIL EKIPPDSEAT LVLVGRADFL EQPVLGFVRL QEAAELEAVE LPVP Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 69%.
Specifications
Specifications
| Antigen | SLC4A1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SLC4A1 |
| Gene Accession No. | P02730 |
| Gene Alias | AE 1; AE1; anion exchange protein 1; anion exchanger 1; anion exchanger-1; band 3 anion transport protein; BND3; CAMP; CD233; CHC; DI; Diego blood group; EMPB3; EPB3; erythrocyte membrane protein band 3; erythroid anion exchange protein; FR; Froese blood group; l11Jus51; MEB3; MGC116750; MGC116753; MGC126619; MGC126623; RTA1A; SAO; Slc4a1; solute carrier family 4 (anion exchanger), member 1; solute carrier family 4 (anion exchanger), member 1 (Diego blood group); solute carrier family 4 member 1; solute carrier family 4 member 1 (Diego blood group); solute carrier family 4, anion exchanger, member 1 (erythrocyte membrane protein band 3, Diego blood group); solute carrier family 4, anion exchanger, number 1; Solute carrier family 4, member 1, anion exchange protein 1 (kidney band 3); SPH4; SW; Swann blood group; Waldner blood group; WD; WD1; WR; Wright blood group; ZNF828 |
| Gene Symbols | SLC4A1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.