Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC4A11 Antibody, Novus Biologicals™
SDP

Catalog No. NBP146156 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP146156 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP146156 Supplier Novus Biologicals Supplier No. NBP146156
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

SLC4A11 Polyclonal specifically detects SLC4A11 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen SLC4A11
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.3-1 ug/ml, Immunohistochemistry-Paraffin 3.75 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_114423.1
Gene Alias Bicarbonate transporter-related protein 1, BTR1bicarbonate transporter related protein 1, CDPD, CHED2, corneal endothelial dystrophy 2 (autosomal recessive), dJ794I6.2, FECD4, MGC126418, MGC126419, NaBC1, sodium bicarbonate transporter-like protein 11, Sodium borate cotransporter 1, sodium-coupled borate cotransporter 1, Solute carrier family 4 member 11, solute carrier family 4, sodium bicarbonate transporter-like, member 11, solute carrier family 4, sodium borate transporter, member 11
Gene Symbols SLC4A11
Host Species Goat
Immunogen Peptide with sequence C-HTSRKYLKLKNFKEE corresponding to internal region according to NP_114423.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 83959
Test Specificity This antibody recognizes and epitope within the internal region. Cross reacts with Cross reacts with Human, Mouse and Rat. Expected from sequence similarity: Dog and Cow.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.