Learn More
Invitrogen™ SLC4A11 Polyclonal Antibody

Description
This antibody is predicted to react with bovine and canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
Specifications
Specifications
| Antigen | SLC4A11 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | SLC4A11 |
| Gene Accession No. | Q8NBS3 |
| Gene Alias | AI503023; bicarbonate transporter related protein 1; bicarbonate transporter-related protein 1; BTR1; CDPD; CDPD1; CHED; CHED2; dJ794I6.2; MGC126418; MGC126419; NABC1; OTTHUMP00000030097; Slc4a11; Sodium bicarbonate transporter-like protein 11; Sodium borate cotransporter 1; sodium-coupled borate cotransporter 1; solute carrier family 4 member 11; solute carrier family 4, sodium bicarbonate transporter-like, member 11; solute carrier family 4, sodium borate transporter, member 11 |
| Gene Symbols | SLC4A11 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.