Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC4A11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101889
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101889 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101889 Supplier Invitrogen™ Supplier No. PA5101889
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SLC4A11.

This gene encodes a voltage-regulated, electrogenic sodium-coupled borate cotransporter that is essential for borate homeostasis, cell growth and cell proliferation. Mutations in this gene have been associated with a number of endothelial corneal dystrophies including recessive corneal endothelial dystrophy 2, corneal dystrophy and perceptive deafness, and Fuchs endothelial corneal dystrophy. Multiple transcript variants encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC4A11
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SLC4A11
Gene Accession No. A2AJN7, Q8NBS3
Gene Alias AI503023; bicarbonate transporter related protein 1; bicarbonate transporter-related protein 1; BTR1; CDPD; CDPD1; CHED; CHED2; dJ794I6.2; MGC126418; MGC126419; NABC1; OTTHUMP00000030097; Slc4a11; Sodium bicarbonate transporter-like protein 11; Sodium borate cotransporter 1; sodium-coupled borate cotransporter 1; solute carrier family 4 member 11; solute carrier family 4, sodium bicarbonate transporter-like, member 11; solute carrier family 4, sodium borate transporter, member 11
Gene Symbols SLC4A11
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLC4A11(Accession Q8NBS3), corresponding to amino acid residues E293-G343.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 269356, 83959
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.