Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC4A1AP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531057
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531057 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531057 Supplier Invitrogen™ Supplier No. PA531057
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (91%), Rat (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

SLC4A1AP gene ontology (GO) annotation includes protein binding and plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC4A1AP
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.76 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene SLC4A1AP
Gene Accession No. Q9BWU0
Gene Alias HLC3; HLC-3; Human lung cancer oncogene 3 protein; kanadaptin; Kidney anion exchanger adapter protein; kidney anion exchanger adaptor protein; lung cancer oncogene 3; SLC4A1AP; solute carrier family 4 (anion exchanger), member 1, adapter protein; solute carrier family 4 (anion exchanger), member 1, adaptor protein; solute carrier family 4 anion exchanger member 1 adapter protein; solute carrier family 4 member 1 adaptor protein
Gene Symbols SLC4A1AP
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SLC4A1AP. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22950
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.