Learn More
Invitrogen™ SLC4A1AP Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Supplier:  Invitrogen™ PA550995
Catalog No. PA550995
Only null left
Add to Cart
Description
Description
SLC4A1AP Polyclonal Antibody detects endogenous levels of SLC4A1AP protein.
SLC4A1AP gene ontology (GO) annotation includes protein binding and plasma membrane.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SLC4A1AP
Polyclonal
Unconjugated
SLC4A1AP
HLC3; HLC-3; Human lung cancer oncogene 3 protein; kanadaptin; Kidney anion exchanger adapter protein; kidney anion exchanger adaptor protein; lung cancer oncogene 3; SLC4A1AP; solute carrier family 4 (anion exchanger), member 1, adapter protein; solute carrier family 4 (anion exchanger), member 1, adaptor protein; solute carrier family 4 anion exchanger member 1 adapter protein; solute carrier family 4 member 1 adaptor protein
Rabbit
Antigen affinity chromatography
RUO
20534, 22950
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4
Q9BWU0
SLC4A1AP
Synthesized peptide derived from the Internal region of human Kanadaptin (aa 421-470).
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.