Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC4A1AP Polyclonal Antibody
SLC4A1AP Polyclonal Antibody
Supplier: Thermo Scientific PA557765
Description
SLC4A1AP Polyclonal Antibody for ICC/IF, IHC

Specifications
SLC4A1AP | |
Polyclonal | |
Unconjugated | |
SLC4A1AP | |
HLC3; HLC-3; Human lung cancer oncogene 3 protein; kanadaptin; Kidney anion exchanger adapter protein; kidney anion exchanger adaptor protein; lung cancer oncogene 3; SLC4A1AP; solute carrier family 4 (anion exchanger), member 1, adapter protein; solute carrier family 4 (anion exchanger), member 1, adaptor protein; solute carrier family 4 anion exchanger member 1 adapter protein; solute carrier family 4 member 1 adaptor protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22950 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BWU0 | |
SLC4A1AP | |
Recombinant protein corresponding to Human SLC4A1AP. Recombinant protein control fragment (Product #RP-102731). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction