Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC4A1AP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115613
Description
Antibody detects endogenous levels of total NADAP.
SLC4A1AP gene ontology (GO) annotation includes protein binding and plasma membrane.
Specifications
SLC4A1AP | |
Polyclonal | |
Unconjugated | |
SLC4A1AP | |
HLC3; HLC-3; Human lung cancer oncogene 3 protein; kanadaptin; Kidney anion exchanger adapter protein; kidney anion exchanger adaptor protein; lung cancer oncogene 3; SLC4A1AP; solute carrier family 4 (anion exchanger), member 1, adapter protein; solute carrier family 4 (anion exchanger), member 1, adaptor protein; solute carrier family 4 anion exchanger member 1 adapter protein; solute carrier family 4 member 1 adaptor protein | |
Rabbit | |
Affinity chromatography | |
RUO | |
20534, 22950 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q9BWU0 | |
SLC4A1AP | |
A synthesized peptide derived from human NADAP, corresponding to a region within C-terminal amino acids. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction