Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC4A2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103606
Description
Antibody detects endogenous levels of total AE2.
Plasma membrane anion exchange protein of wide distribution.
Specifications
SLC4A2 | |
Polyclonal | |
Unconjugated | |
SLC4A2 | |
AE 2; AE2; Aep2; anion exchange protein 2; anion exchanger 2; anion exchanger 2 type a; anion exchanger 2 type b1; anion exchanger 2 type b2; B3RP; B3rp2; B3RP-2; Band 3-related protein; band 3-related protein 2; BND3L; EPB3L1; Erythrocyte band 3; erythrocyte membrane protein band 3-like 1; HKB3; MPB3L; NBND3; non-erythroid band 3-like protein; Slc4a2; solute carrier family 4 (anion exchanger), member 2; solute carrier family 4 member 2; Solute carrier family 4, member 2, anion exchange protein 2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
20535, 24780, 6522 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P04920, P13808, P23347 | |
SLC4A2 | |
A synthesized peptide derived from human SLC4A2(Accession P04920), corresponding to amino acid residues N868-L918. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction