Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC4A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530172
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530172 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530172 Supplier Invitrogen™ Supplier No. PA530172
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549. Predicted reactivity: Mouse (97%), Rat (96%), Rabbit (96%), Chicken (82%), Bovine (98%), Guinea pig (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Plasma membrane anion exchange protein of wide distribution.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC4A2
Applications Western Blot
Classification Polyclonal
Concentration 0.74 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene SLC4A2
Gene Accession No. P04920
Gene Alias AE 2; AE2; Aep2; anion exchange protein 2; anion exchanger 2; anion exchanger 2 type a; anion exchanger 2 type b1; anion exchanger 2 type b2; B3RP; B3rp2; B3RP-2; Band 3-related protein; band 3-related protein 2; BND3L; EPB3L1; Erythrocyte band 3; erythrocyte membrane protein band 3-like 1; HKB3; MPB3L; NBND3; non-erythroid band 3-like protein; Slc4a2; solute carrier family 4 (anion exchanger), member 2; solute carrier family 4 member 2; Solute carrier family 4, member 2, anion exchange protein 2
Gene Symbols SLC4A2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the Intracellular domain of human SLC4A2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6522
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.