Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC4A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA551351
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA551351 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA551351 Supplier Invitrogen™ Supplier No. PA551351
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total SLC4A3 protein.

Plasma membrane anion exchange protein of wide distribution. Mediates at least a part of the Cl-/HCO3- exchange in cardiac myocytes. Both BAE3 and CAE3 forms transport Cl-.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC4A3
Applications Western Blot
Classification Polyclonal
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene SLC4A3
Gene Accession No. P16283, P23348, P48751
Gene Alias A930038D23Rik; AE 3; Ae3; Aep3; anion exchange protein 3; anion exchanger 3; anion exchanger 3 cardiac isoform; Anion exchanger 3, neuronal; B3rp3; B3RP-3; Band 3-related protein 3; CAE3/BAE3; Cardiac/brain band 3-like protein; neuronal band 3-like protein; SLC2C; Slc4a3; solute carrier family 4 (anion exchanger), member 3; solute carrier family 4 member 3; Solute carrier family 4, member 3, anion exchange protein 3
Gene Symbols SLC4A3
Host Species Rabbit
Immunogen Synthetic peptide of human SLC4A3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20536, 24781, 6508
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.