Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC4A4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$343.50 - $573.00
Specifications
Antigen | SLC4A4 |
---|---|
Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SLC4A4 Polyclonal antibody specifically detects SLC4A4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
SLC4A4 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Endocrinology, Neuroscience, Signal Transduction | |
PBS, pH 7.2, 40% glycerol | |
8671 | |
IgG | |
Affinity purified |
Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
electrogenic sodium bicarbonate cotransporter 1, hhNMC, HNBC1, KNBC, kNBC1, Na(+)/HCO3(-) cotransporter, NBC, NBC1DKFZp781H1314, NBC2, NBCE1, pNBC, SLC4A5, Sodium bicarbonate cotransporter, sodium bicarbonate cotransporter 1 (sodium bicarbonate cotransporter, kidney;sodium bicarbonate cotransporter, pancreas), Solute carrier family 4 member 4, solute carrier family 4, sodium bicarbonate cotransporter, member 4, solute carrier family 4, sodium bicarbonate cotransporter, member 4, brain type, solute carrier family 4, sodium bicarbonate cotransporter, member 5 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: DIGKTVSSASRMFTNPDNGSPAMTHRNLTSSSLNDISDKPEKDQLKNKF | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title