Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC4A5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583342
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583342 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583342 Supplier Invitrogen™ Supplier No. PA583342
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SQHDLAWIDN ILPEKEKKET DKKRKRKKGA HEDCDEEPQF PPPSVIKIPM ESVQSDPQNG IHCIARKRSS SWSYSL Highest antigen sequence indentity to the following orthologs: Mouse - 77%, Rat - 76%.

This gene encodes a member of the sodium bicarbonate cotransporter (NBC) family, part of the bicarbonate transporter superfamily. Sodium bicarbonate cotransporters are involved in intracellular pH regulation and electroneural or electrogenic sodium bicarbonate transport. This protein is thought to be an integral membrane protein. Multiple transcript variants encoding different isoforms have been found for this gene, but the biological validity of some variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC4A5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC4A5
Gene Accession No. Q9BY07
Gene Alias C330006M16; C330016K18Rik; electrogenic sodium bicarbonate cotransporter 4; electrogenic sodium bicarbonate cotransporter NBC4c; NBC4; NBCe2; RGD:1303009}; Slc4a5; slc4a5 {ECO:0000312; sodium bicarbonate transporter 4; solute carrier family 4 (sodium bicarbonate cotransporter), member 5; solute carrier family 4 member 5; solute carrier family 4, sodium bicarbonate cotransporter, member 5
Gene Symbols SLC4A5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC4A5. Recombinant protein control fragment (Product #RP-96222).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57835
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.