Learn More
Invitrogen™ SLC4A5 Polyclonal Antibody

Description
Immunogen sequence: DFIFSQHDLA WIDNILPEKE KKETDKKRKR KKGAHEDCDE EPQFPPPSVI KIPMESVQSD PQNGIHCIAR KRSSSWSYSL; Positive Samples: LO2, U-87MG, SW480, SGC-7901, Mouse kidney; Cellular Location: Apical cell membrane, Multi-pass membrane protein.
Specifications
Specifications
| Antigen | SLC4A5 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.45 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SLC4A5 |
| Gene Accession No. | Q6RI88, Q9BY07 |
| Gene Alias | C330006M16; C330016K18Rik; electrogenic sodium bicarbonate cotransporter 4; electrogenic sodium bicarbonate cotransporter NBC4c; NBC4; NBCe2; RGD:1303009}; Slc4a5; slc4a5 {ECO:0000312; sodium bicarbonate transporter 4; solute carrier family 4 (sodium bicarbonate cotransporter), member 5; solute carrier family 4 member 5; solute carrier family 4, sodium bicarbonate cotransporter, member 5 |
| Gene Symbols | SLC4A5 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.