Learn More
Invitrogen™ SLC4A7 Polyclonal Antibody

Description
Positive Samples: Mouse liver Immunogen sequence: KKEKEEAERM LQDDDDTVHL PFEGGSLLQI PVKALKYSPD KPVSVKISFE DEPRKKYVDA ETSL.
Specifications
Specifications
| Antigen | SLC4A7 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 2.27 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 |
| Gene | SLC4A7 |
| Gene Accession No. | Q8BTY2, Q9Y6M7 |
| Gene Alias | bicarbonate transporter; BT; E430014N10Rik; Electroneutral Na/HCO(3) cotransporter; electroneutral sodium bicarbonate cotransporter 1; Na-bicarbonate cotransporter; Nbc; NBC2; NBC2B; NBC3; NBC-like protein; Nbcn1; NBCn1-G; SBC2; SLC4A6; Slc4a7; Sodium bicarbonate cotransporter 2; sodium bicarbonate cotransporter 2b; Sodium bicarbonate cotransporter 3; sodium bicarbonate cotransporter NBCn1-A; sodium bicarbonate cotransporter NBCn1-B; sodium bicarbonate cotransporter NBCn1-c; sodium bicarbonate cotransporter NBCn1-D; sodium bicarbonate cotransporter NBCn1-H; solute carrier family 4 member 7; solute carrier family 4, sodium bicarbonate cotransporter, member 7 |
| Gene Symbols | SLC4A7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.