Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC4A8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101904
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA5101904 100 μL
1 options

Catalog No. PIPA5101904

Supplier: Invitrogen™ PA5101904

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SLC4A8/10.

Mediates electroneutral sodium- and carbonate-dependent choride-HCO3(-) exchange with a Na(+):HCO3(-) stoichiometry of 2:1. Plays a major role in pH regulation in neurons. May be involved in cell pH regulation by transporting HCO3(-) from blood to cell. Enhanced expression in severe acid stress could be important for cell survival by mediating the influx of HCO3(-) into the cells. Also mediates lithium-dependent HCO3(-) cotransport. May be regulated by osmolarity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC4A8
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Slc4a8
Gene Accession No. Q2Y0W8, Q6RVG2, Q8JZR6
Gene Alias AW493845; C230026C11; Electroneutral Na(+)-driven Cl-HCO3 exchanger; electroneutral Na+-driven Cl-HCO3 exchanger; Electroneutral sodium bicarbonate exchanger 1; KIAA0739; k-NBC3; kNBC-3; Na-driven Cl-HCO3 exchanger NDCBE1-A; Na-driven Cl-HCO3 exchanger NDCBE1-B; Na-driven Cl-HCO3 exchanger NDCBE1-C; NBC; NBC3; NDCBE; NDCBE1; RGD:735164}; Slc4a8; slc4a8 {ECO:0000312; sodium bicarbonate cotransporter isoform 3 kNBC-3; solute carrier family 4 (anion exchanger), member 8; solute carrier family 4 member 8; solute carrier family 4, sodium bicarbonate cotransporter, member 8
Gene Symbols Slc4a8
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLC4A8(Accession Q2Y0W8), corresponding to amino acid residues F1040-S1090.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 315311, 59033, 9498
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.