Learn More
Invitrogen™ SLC4A10 Polyclonal Antibody

Description
Immunogen sequence: FELSEAYPIN MHNDLELLTQ YSCNCVEPHN PSNGTLKEWR ESNISASDII WENLT Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 91%.
Specifications
Specifications
| Antigen | SLC4A10 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SLC4A10 |
| Gene Accession No. | Q6U841 |
| Gene Alias | NBCN2; NCBE; Sodium-Driven Chloride Bicarbonate Exchanger |
| Gene Symbols | SLC4A10 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.