Learn More
Invitrogen™ SLC4A9 Polyclonal Antibody

Description
This antibody detects endogenous protein at a molecular weight of 108 kDa. Purity is >95% by SDS-PAGE.
Specifications
Specifications
| Antigen | SLC4A9 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide |
| Gene | SLC4A9 |
| Gene Accession No. | Q8K4V2, Q96Q91 |
| Gene Alias | AE 4; AE4; Anion exchange protein 4; anion exchanger 4; D630003B07; D630024F24Rik; SBC5; SLC4A9; Sodium bicarbonate cotransporter 5; solute carrier family 4 member 9; solute carrier family 4, sodium bicarbonate cotransporter, member 9 |
| Gene Symbols | SLC4A9 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.