Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC50A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553721
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553721 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553721 Supplier Invitrogen™ Supplier No. PA553721
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RMTRSVDNVQ FLPFLTTEVN NLGWLSYGAL KGDG Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 85%.

SLC50A1 gene ontology annotations related to this gene include carbohydrate transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC50A1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC50A1
Gene Accession No. Q9BRV3
Gene Alias HsSWEET1; LOW QUALITY PROTEIN: sugar transporter SWEET1; MmSWEET1; probable sugar transporter RAG1AP1; RAG1-activating protein 1; RAG1AP1; recombination activating gene 1 activating protein 1; recombination activating gene 1 gene activation; Rga; RGD1308478; RP11-540D14.5; RZPDo834D038D; SCP; SLC50A1; slv; solute carrier family 50 (sugar efflux transporter), member 1; solute carrier family 50 (sugar transporter), member 1; solute carrier family 50 member 1; Stromal cell protein; sugar transporter SWEET1; SWEET1
Gene Symbols SLC50A1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC50A1. Recombinant protein control fragment (Product #RP-91097).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55974
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.