Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC52A1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5102239
Description
Antibody detects endogenous levels of total PEVR2.
Biological redox reactions require electron donors and acceptor. Vitamin B2 is the source for the flavin in flavin adenine dinucleotide (FAD) and flavin mononucleotide (FMN) which are common redox reagents. This gene encodes a member of the riboflavin (vitamin B2) transporter family. Haploinsufficiency of this protein can cause maternal riboflavin deficiency. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq, Jan 2013].
Specifications
SLC52A1 | |
Polyclonal | |
Unconjugated | |
SLC52A1 | |
G protein-coupled receptor 172B; GPCR42; GPR172B; G-protein coupled receptor GPCR42; hRFT1; PAR2; PERV-A receptor 2; PEVR2; porcine endogenous retrovirus A receptor 2; Protein GPR172B; RBFVD; RFT1; RFT1sv; RFVT1; Riboflavin transporter 1; SLC52A1; solute carrier family 52 (riboflavin transporter), member 1; solute carrier family 52 member 1; solute carrier family 52, riboflavin transporter, member 1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
55065 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9NWF4 | |
SLC52A1 | |
A synthesized peptide derived from human SLC52A1(Accession Q9NWF4), corresponding to amino acid residues E243-T293. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction