Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC52A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102238
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102238 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102238 Supplier Invitrogen™ Supplier No. PA5102238
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PEVR1.

This gene encodes a membrane protein which belongs to the riboflavin transporter family. In humans, riboflavin must be obtained by intestinal absorption because it cannot be synthesized by the body. The water-soluble vitamin riboflavin is processed to the coenzymes flavin mononucleotide (FMN) and flavin adenine dinucleotide (FAD) which then act as intermediaries in many cellular metabolic reactions. Paralogous members of the riboflavin transporter gene family are located on chromosomes 17 and 20. Unlike other members of this family, this gene has higher expression in brain tissue than small intestine. Alternative splicing of this gene results in multiple transcript variants encoding the same protein. Mutations in this gene have been associated with Brown-Vialetto-Van Laere syndrome 2 - an autosomal recessive progressive neurologic disorder characterized by deafness, bulbar dysfunction, and axial and limb hypotonia. [provided by RefSeq, Jul 2012].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC52A2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SLC52A2
Gene Accession No. Q9HAB3
Gene Alias 2010003P03Rik; BVVLS2; D15Ertd747e; G protein-coupled receptor 172A; G protein-coupled receptor 172B; GPCR41; GPCR42; Gpr172a; Gpr172b; hRFT3; mRFT1; PAR1; PAR2; PERV-A receptor 1; PERV-A receptor 2 homolog; PEVR1; Porcine endogenous retrovirus A receptor 1; porcine endogenous retrovirus A receptor 2; Porcine endogenous retrovirus A receptor 2 homolog; Protein GPR172A; Protein GPR172B; putative G-protein coupled receptor GPCR41; Rft1; RFT3; RFVT2; RGD1560410; riboflavin transporter 1; Riboflavin transporter 3; rRFT1; Slc52a2; solute carrier family 52 (riboflavin transporter), member 2; solute carrier family 52 member 2; solute carrier family 52, riboflavin transporter, member 2; solute carrier protein 52, member 2
Gene Symbols SLC52A2
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLC52A2(Accession Q9HAB3), corresponding to amino acid residues Y50-H100.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79581
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.