Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC5A10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533039
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533039 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533039 Supplier Thermo Scientific Supplier No. PA533039
Add to Cart
Add to Cart

SLC5A10 Polyclonal Antibody

SLC5A10 Polyclonal Antibody for IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC5A10
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.85 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene SLC5A10
Gene Accession No. A0PJK1
Gene Alias Na(+)/glucose cotransporter 5; SGLT5; SGLT-5; SLC5A10; Sodium/glucose cotransporter 5; solute carrier family 5 (sodium/glucose cotransporter), member 10; solute carrier family 5 (sodium/sugar cotransporter), member 10; solute carrier family 5 member 10
Gene Symbols SLC5A10
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from cytoplasmic domain of human SLC5A10.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 125206
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.