Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC5A10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571397
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571397 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571397 Supplier Invitrogen™ Supplier No. PA571397
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC5A10 Polyclonal Antibody for Western Blot

SGLT5 is a 62 kDa sodium/glucose cotransporter whose expression is restricted to kidney, and is thought to have a role in renal function. Multiple splicing transcripts exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC5A10
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC5A10
Gene Accession No. A0PJK1
Gene Alias Na(+)/glucose cotransporter 5; SGLT5; SGLT-5; SLC5A10; Sodium/glucose cotransporter 5; solute carrier family 5 (sodium/glucose cotransporter), member 10; solute carrier family 5 (sodium/sugar cotransporter), member 10; solute carrier family 5 member 10
Gene Symbols SLC5A10
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human SLC5A10.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 125206
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.