Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC5A11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533040
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533040 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533040 Supplier Invitrogen™ Supplier No. PA533040
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset (100%); Rabbit, Horse, Pig (94%); Rat, Elephant (88%); Mouse, Dog, Bovine, Hamster, Opossum, Turkey, Chicken (82%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

Cotransporters, such as SLC5A11, represent a major class of proteins that make use of ion gradients to drive active transport for the cellular accumulation of nutrients, neurotransmitters, osmolytes, and ions Roll et al. (2002).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC5A11
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene SLC5A11
Gene Accession No. Q8WWX8
Gene Alias 2010013B02Rik; homolog of rabbit KST1; hypothetical protein LOC539084; KST1; Na(+)/myo-inositol cotransporter 2; na+/myo-inositol cotransporter 2; Na+/myo-inositol cotransporter SMIT2; putative sodium-coupled cotransporter RKST1; R SGLT6; RGD:621410}; RKST1; RKST2; SGLT6; Slc5a10; slc5a11; slc5a11 {ECO:0000312; SLGTX; SMIT2; sodium/glucose cotransporter KST1; Sodium/myo-inositol cotransporter 2; sodium/myo-inositol transporter 2; sodium-cotransporter rkST1; sodium-coupled cotransporter KST1; sodium-dependent glucose cotransporter; sodium-glucose cotransporter-like protein; solute carrier family 5 (sodium/glucose cotransporter), member 10; solute carrier family 5 (sodium/glucose cotransporter), member 11; solute carrier family 5 (sodium/inositol cotransporter), member 11; solute carrier family 5 member 11; zgc:92080
Gene Symbols SLC5A11
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from extracellular domain of human SLC5A11.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115584
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.