Learn More
Invitrogen™ SLC5A11 Polyclonal Antibody

Description
Immunogen sequence: SWFTEPPSKE MVSHLTWFTR HDPVVQKEQA PPAAPLSLTL SQNGMPEASS SSSVQFEMVQ ENTSKTHSCD MTPKQSKVVK AILWLCGIQE KGKEELPARA E; Positive Samples: 293T, U-87MG, Mouse kidney, Rat brain; Cellular Location: Membrane, Multi-pass membrane protein.
Specifications
Specifications
| Antigen | SLC5A11 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 3.07 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | SLC5A11 |
| Gene Accession No. | Q8K0E3, Q8WWX8, Q9Z1F2 |
| Gene Alias | 2010013B02Rik; homolog of rabbit KST1; hypothetical protein LOC539084; KST1; Na(+)/myo-inositol cotransporter 2; na+/myo-inositol cotransporter 2; Na+/myo-inositol cotransporter SMIT2; putative sodium-coupled cotransporter RKST1; R SGLT6; RGD:621410}; RKST1; RKST2; SGLT6; Slc5a10; slc5a11; slc5a11 {ECO:0000312; SLGTX; SMIT2; sodium/glucose cotransporter KST1; Sodium/myo-inositol cotransporter 2; sodium/myo-inositol transporter 2; sodium-cotransporter rkST1; sodium-coupled cotransporter KST1; sodium-dependent glucose cotransporter; sodium-glucose cotransporter-like protein; solute carrier family 5 (sodium/glucose cotransporter), member 10; solute carrier family 5 (sodium/glucose cotransporter), member 11; solute carrier family 5 (sodium/inositol cotransporter), member 11; solute carrier family 5 member 11; zgc:92080 |
| Gene Symbols | SLC5A11 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.