Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC5A12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563910
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563910 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563910 Supplier Invitrogen™ Supplier No. PA563910
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QGSTHAGGFH NVLEQSTNGS RLHIFDFDVD PLRRH Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 80%.

Normal blood lactate is maintained at about 1.5 mM, and little filtered lactate is excreted in urine. Reabsorption of lactate is mediated by the low-affinity Na(+)-coupled lactate transporter SLC5A12 in the initial part of the proximal tubule and by the high-affinity Na(+)-coupled lactate transporter SLC5A8 (MIM 608044) in the distal proximal tubule (Gopal et al., 2007 [PubMed 17692818]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC5A12
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC5A12
Gene Accession No. Q1EHB4
Gene Alias electroneutral sodium monocarboxylate cotransporter; Low-affinity sodium-lactate cotransporter; SLC5A12; SMCT2; Sodium-coupled monocarboxylate transporter 2; sodium-iodide related cotransporter; solute carrier family 5 (sodium/glucose cotransporter), member 12; solute carrier family 5 (sodium/monocarboxylate cotransporter), member 12; solute carrier family 5 member 12
Gene Symbols SLC5A12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC5A12. Recombinant protein control fragment (Product #RP-104133).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 159963
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.