Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC5A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01674437
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-437 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-437 Supplier Invitrogen™ Supplier No. PA19554
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

PA1-9554 detects SLC5A3 in human samples. PA1-9554 has been successfully used in Western blot procedures. PA1-9554 immunogen is a purified recombinant SLC5A3 corresponding to the chromosome 21 gene (Human).

Prevents intracellular accumulation of high concentrations of myo-inositol (an osmolyte) that result in impairment of cellular function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC5A3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SLC5A3
Gene Accession No. P53794
Gene Alias AA623876; BCW2; BCW2 protein; BF642829; Na(+)/myo-inositol cotransporter; Slc5a3; SMIT; Smit1; SMIT2; sodium myo-inositol transporter 1; sodium/myo-inositol cotransporter; sodium/myo-inositol cotransporter 1; sodium/myo-inositol transporter 1; solute carrier family 5 (inositol transporters), member 3; solute carrier family 5 (sodium/myo-inositol cotransporter), member 3; solute carrier family 5 member 3; solute carrier family 5, member 3; solute carrier family 5, member alpha 3
Gene Symbols SLC5A3
Host Species Chicken
Immunogen Purified recombinant SLC5A3 corresponding to the chromosome 21 gene (Human).
Purification Method IgY fraction
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6526
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.