Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC5A3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534211
Description
Percent identity with other species by BLAST analysis: Baboon, Monkey, Marmoset (100%) Human, Galago, Elephant, Panda, Bat, Bovine, Dog, Rabbit, Pig (94%) Rat (88%).
Prevents intracellular accumulation of high concentrations of myo-inositol (an osmolyte) that result in impairment of cellular function.
Specifications
SLC5A3 | |
Polyclonal | |
Unconjugated | |
SLC5A3 | |
AA623876; BCW2; BCW2 protein; BF642829; Na(+)/myo-inositol cotransporter; Slc5a3; SMIT; Smit1; SMIT2; sodium myo-inositol transporter 1; sodium/myo-inositol cotransporter; sodium/myo-inositol cotransporter 1; sodium/myo-inositol transporter 1; solute carrier family 5 (inositol transporters), member 3; solute carrier family 5 (sodium/myo-inositol cotransporter), member 3; solute carrier family 5 member 3; solute carrier family 5, member 3; solute carrier family 5, member alpha 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6526 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P53794 | |
SLC5A3 | |
Synthetic 17 amino acid peptide from internal region of human SLC5A3. | |
50 μg | |
Primary | |
Human, Monkey | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction