Learn More
Invitrogen™ SLC5A3 Polyclonal Antibody

Description
SMIT2 / SLC5A3 Polyclonal Antibody detects endogenous levels of SMIT2 / SLC5A3 protein.
Specifications
Specifications
| Antigen | SLC5A3 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | SLC5A3 |
| Gene Accession No. | P53794, Q9JKZ2 |
| Gene Alias | AA623876; BCW2; BCW2 protein; BF642829; Na(+)/myo-inositol cotransporter; Slc5a3; SMIT; Smit1; SMIT2; sodium myo-inositol transporter 1; sodium/myo-inositol cotransporter; sodium/myo-inositol cotransporter 1; sodium/myo-inositol transporter 1; solute carrier family 5 (inositol transporters), member 3; solute carrier family 5 (sodium/myo-inositol cotransporter), member 3; solute carrier family 5 member 3; solute carrier family 5, member 3; solute carrier family 5, member alpha 3 |
| Gene Symbols | SLC5A3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.