Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC5A4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533044
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533044 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533044 Supplier Invitrogen™ Supplier No. PA533044
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla (100%); Gibbon, Baboon, Monkey (94%); Marmoset (81%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

Has electrogenic activity in response to glucose, and may function as a glucose sensor (PubMed:13130073, PubMed:17110502, PubMed:20421923, PubMed:22766068). Mediates influx of sodium ions into the cell but does not transport sugars (PubMed:13130073, PubMed:22766068). Also potently activated by imino sugars such as deoxynojirimycin (DNJ) (PubMed:17110502, PubMed:20421923, PubMed:22766068). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC5A4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.88 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene SLC5A4
Gene Accession No. Q9NY91
Gene Alias DJ90G24.4; low affinity sodium glucose cotransporter; low affinity sodium-glucose cotransporter; Na(+)/glucose cotransporter 3; SAAT1; SGLT3; SLC5A4; Slc5a4a; sodium transporter; sodium/glucose cotransporter 3; solute carrier family 5 (glucose activated ion channel), member 4; solute carrier family 5 (low affinity glucose cotransporter), member 4; solute carrier family 5 (low affinity glucose cotransporter), member 4a; solute carrier family 5 (neutral amino acid transporters, system A), member 4; solute carrier family 5 member 4; solute carrier family 5, member 4; solute carrier family 5, member 4a
Gene Symbols SLC5A4
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from C-terminus of human SLC5A4.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6527
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.