Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC5A5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113168
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113168 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113168 Supplier Invitrogen™ Supplier No. PA5113168
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC5A5 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Human Sodium Iodide Symporter (hNIS) is responsible for iodide concentrating ability within thyroid follicular cells. It is a membrane bound glycoprotein with 13 membrane spanning domains and 14 extramembranous domains. It may represent an autoantigen in thyroid.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC5A5
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SLC5A5
Gene Accession No. Q63008, Q92911
Gene Alias Na(+)/I(-) cotransporter; Na(+)/I(-) symporter; Na(+)/I(-)-symporter; Na+/I- symporter; Nis; SLC5A5; sodium iodide symporter; Sodium/iodide cotransporter; sodium-iodide symporter; solute carrier family 5 (sodium iodide symporter), member 5; solute carrier family 5 (sodium/iodide cotransporter), member 5; solute carrier family 5 member 5; solute carrier family 5, member 5; TDH1
Gene Symbols SLC5A5
Host Species Rabbit
Immunogen Recombinant Human Sodium/iodide cotransporter protein (547-643AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114613, 6528
Target Species Human, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.