Learn More
SLC5A5/Sodium Iodide Symporter Antibody (14F), DyLight 755, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP259341IR
Catalog No. NBP259341IR
Only null left
Add to Cart
Description
Description
SLC5A5/Sodium Iodide Symporter Monoclonal specifically detects SLC5A5/Sodium Iodide Symporter in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
SLC5A5/Sodium Iodide Symporter
Monoclonal
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
SLC5A5
Mannose binding protein hNIS fusion (AA468-643)
100 μL
6528
Human, Mouse, Rat
Purified
Western Blot, Immunohistochemistry
14F
Western Blot, Immunohistochemistry
Na(+)/I(-) cotransporter, Na(+)/I(-) symporter, NISNa(+)/I(-)-symporter, sodium/iodide cotransporter, Sodium-iodide symporter, solute carrier family 5 (sodium iodide symporter), member 5, Solute carrier family 5 member 5, TDH1
Mouse
Protein G purified
Primary
Detects 97kDa, non-glycosylated version at 68kDa. Other minor bands associated with hNIS at 160kDa, and degradation products at 30 kDa, and 15kDa.
Store at 4C in the dark.
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.