Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC5A5/Sodium Iodide Symporter Antibody (14F), Novus Biologicals™

Mouse Monoclonal Antibody
$560.50
Specifications
Antigen | SLC5A5/Sodium Iodide Symporter |
---|---|
Clone | 14F |
Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Description
SLC5A5/Sodium Iodide Symporter Monoclonal specifically detects SLC5A5/Sodium Iodide Symporter in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Microarray.Specifications
SLC5A5/Sodium Iodide Symporter | |
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
Monoclonal | |
Purified | |
RUO | |
Na(+)/I(-) cotransporter, Na(+)/I(-) symporter, NISNa(+)/I(-)-symporter, sodium/iodide cotransporter, Sodium-iodide symporter, solute carrier family 5 (sodium iodide symporter), member 5, Solute carrier family 5 member 5, TDH1 | |
SLC5A5 | |
IgG1 | |
Protein G purified |
14F | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
6528 | |
Mannose binding protein hNIS fusion (AA468-643) | |
Primary | |
Detects 97kDa, non-glycosylated version at 68kDa. Other minor bands associated with hNIS at 160kDa, and degradation products at 30 kDa, and 15kDa. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title