Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC5A5/Sodium Iodide Symporter Antibody (902320), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB8367E
Description
SLC5A5/Sodium Iodide Symporter Monoclonal antibody specifically detects SLC5A5/Sodium Iodide Symporter in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
SLC5A5/Sodium Iodide Symporter | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
902320 | |
Flow Cytometry, CyTOF-ready | |
Na(+)/I(-) cotransporter, Na(+)/I(-) symporter, NISNa(+)/I(-)-symporter, sodium/iodide cotransporter, Sodium-iodide symporter, solute carrier family 5 (sodium iodide symporter), member 5, Solute carrier family 5 member 5, TDH1 | |
NS0 mouse myeloma cell line transfected with human SLC5A5, Accession # Q92911 | |
0.1 mL | |
ABC Transporters, Cancer, Cellular Signaling, Neuroscience | |
6528 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction