Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC5A6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110390
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110390 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110390 Supplier Invitrogen™ Supplier No. PA5110390
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TVATVTTLMP LTTFSKPTGL QRFYSLSYLW YSAHNSTTVI VVGLIVSLLT GRMRGRSLNP ATIYPVLPKL LSLLPLSCQK RLHCRSYGQD HLDTGLFPEK PRNGVLGDSR DKEAMALDGT AYQGSSSTCI LQETSL.

Transports pantothenate, biotin and lipoate in the presence of sodium. Target protein mediates sodium-dependent transport of pantothenate, biotin, and lipoate; plays a role in intestinal uptake of biotin and in transplacental vitamin transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC5A6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.92 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SLC5A6
Gene Accession No. Q9Y289
Gene Alias E430023I20; Na(+)-dependent multivitamin transporter; Na+-dependent multivitamin transporter; Slc5a6; SMVT; Sodium-dependent multivitamin transporter; solute carrier family 5 (sodium/multivitamin and iodide cotransporter), member 6; solute carrier family 5 (sodium-dependent vitamin transporter), member 6; solute carrier family 5 member 6; solute carrier family 5, member 6
Gene Symbols SLC5A6
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 500 to the C-terminus of human SLC5A6 (NP_066918.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8884
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.