Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC5A7 Monoclonal Antibody (62-2E8)
GREENER_CHOICE

Catalog No. PIMA146409
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA146409 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA146409 Supplier Invitrogen™ Supplier No. MA146409
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

In Western blot a band can be seen at ~65 kDa representing CHT and a band can also be seen at ~45 kDa after deglycosylation with PNGase F. Suggested positive control: antigen standard for SLC5A7 (transient overexpression lysate), rat striatum protein.

Acetylcholine functions as a neurotransmitter in both the central and peripheral nervous systems of all vertebates, and is the principle neurotransmitter used at the neuromuscular junction. Presynaptic synthesis of acetylcholine requires a steady supply of choline, which is acquired by a plasma membrane high affinity choline transporter (CHT).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC5A7
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 62-2E8
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% sodium azide
Gene Slc5a7
Gene Accession No. Q8BGY9, Q9GZV3, Q9JMD7
Gene Alias CHT; CHT1; hCHT; Hemicholinium-3-sensitive choline transporter; high affinity choline transporter 1; high affinity choline transporter 1; LOW QUALITY PROTEIN: high affinity choline transporter 1; high affinity choline transporter; hemicholinium-3-sensitive choline transporter; HMN7A; MGC126299; MGC126300; similar to solute carrier family 5 (choline transporter), member 7; SLC5A7; solute carrier family 5 (choline transporter), member 7; solute carrier family 5 (sodium/choline cotransporter), member 7; solute carrier family 5 member 7; solute carrier family 5, member 7
Gene Symbols Slc5a7
Host Species Mouse
Immunogen Recombinant fusion protein from the C-terminal portion of the human CHT1 protein.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60482, 63993, 85426
Target Species Human, Mouse, Rat, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.