Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC5A7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117124
Description
Positive Samples: C6, SH-SY5Y, U-251MG, SGC-7901, Mouse brain, Rat brain Immunogen sequence: ESGTLPPKLD VFDAVVARHS EENMDKTILV KNENIKLDEL ALVKPRQSMT LSSTFTNKEA FLDVDSSPEG SGTEDNLQ.
Acetylcholine functions as a neurotransmitter in both the central and peripheral nervous systems of all vertebates, and is the principle neurotransmitter used at the neuromuscular junction. Presynaptic synthesis of acetylcholine requires a steady supply of choline, which is acquired by a plasma membrane high affinity choline transporter (CHT).
Specifications
SLC5A7 | |
Polyclonal | |
Unconjugated | |
Slc5a7 | |
CHT; CHT1; hCHT; Hemicholinium-3-sensitive choline transporter; high affinity choline transporter 1; high affinity choline transporter 1; LOW QUALITY PROTEIN: high affinity choline transporter 1; high affinity choline transporter; hemicholinium-3-sensitive choline transporter; HMN7A; MGC126299; MGC126300; similar to solute carrier family 5 (choline transporter), member 7; SLC5A7; solute carrier family 5 (choline transporter), member 7; solute carrier family 5 (sodium/choline cotransporter), member 7; solute carrier family 5 member 7; solute carrier family 5, member 7 | |
Rabbit | |
Affinity chromatography | |
RUO | |
60482, 63993, 85426 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BGY9, Q9GZV3, Q9JMD7 | |
Slc5a7 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 503-580 of human SLC5A7 (NP_0685871). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction