Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC5A8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568431
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568431 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568431 Supplier Invitrogen™ Supplier No. PA568431
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC5A8 Polyclonal Antibody for IHC (P)

SLC5A8 has been shown to transport iodide by a passive mechanism (Rodriguez et al., 2002 ) and monocarboxylates and short-chain fatty acids by a sodium-coupled mechanism (Gopal et al., 2004 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC5A8
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Slc5a8
Gene Accession No. Q8N695
Gene Alias AIT; Apical iodide transporter; apical iodide transporter , putative; electrogenic sodium monocarboxylate cotransporter; RGD1564146; Slc5a8; SMCT; SMCT1; Sodium iodide-related cotransporter; sodium-coupled monocarboxylate transporter 1; solute carrier family 5 (iodide transporter), member 8; solute carrier family 5 (sodium/monocarboxylate cotransporter), member 8; solute carrier family 5 member 8
Gene Symbols Slc5a8
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues near the C terminal of human SLC5A8.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 160728
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.