Learn More
Invitrogen™ SLC5A9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533047
Description
Percent identity with other species by BLAST analysis: Human, Gorilla (100%); Monkey (94%); Gibbon, Marmoset, Bovine (89%); Mouse (83%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
Involved in sodium-dependent transport of D-mannose, D-glucose and D-fructose.
Specifications
SLC5A9 | |
Polyclonal | |
Unconjugated | |
SLC5A9 | |
hSGLT4; Na(+)/glucose cotransporter 4; SGLT4; SLC5A9; Sodium/glucose cotransporter 4; solute carrier family 5 (sodium/glucose cotransporter), member 9; solute carrier family 5 (sodium/sugar cotransporter), member 9; solute carrier family 5 member 9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
200010 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q2M3M2 | |
SLC5A9 | |
Synthetic 18 amino acid peptide from C-terminus of human SLC5A9. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.