Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC5A9 Polyclonal Antibody

SLC5A9 Polyclonal Antibody
Supplier: Thermo Scientific PA533049
Description
SLC5A9 Polyclonal Antibody for IHC (P)
Specifications
| SLC5A9 | |
| Polyclonal | |
| Unconjugated | |
| SLC5A9 | |
| hSGLT4; Na(+)/glucose cotransporter 4; SGLT4; SLC5A9; Sodium/glucose cotransporter 4; solute carrier family 5 (sodium/glucose cotransporter), member 9; solute carrier family 5 (sodium/sugar cotransporter), member 9; solute carrier family 5 member 9 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 200010 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q2M3M2 | |
| SLC5A9 | |
| Synthetic 17 amino acid peptide from cytoplasmic domain of human SLC5A9. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction