Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC5A9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534212
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534212 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534212 Supplier Invitrogen™ Supplier No. PA534212
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Orangutan, Gibbon, Monkey, Marmoset (100%); Elephant, Dog (94%); Galago, Panda, Bovine, Horse, Pig (88%); Mouse, Bat (82%). Proteinase K treatment is required for antigen retrieval.

Involved in sodium-dependent transport of D-mannose, D-glucose and D-fructose.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC5A9
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene SLC5A9
Gene Accession No. Q2M3M2
Gene Alias hSGLT4; Na(+)/glucose cotransporter 4; SGLT4; SLC5A9; Sodium/glucose cotransporter 4; solute carrier family 5 (sodium/glucose cotransporter), member 9; solute carrier family 5 (sodium/sugar cotransporter), member 9; solute carrier family 5 member 9
Gene Symbols SLC5A9
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from internal region of human SLC5A9.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200010
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.