Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC5A9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569254
Description
This target displays homology in the following species: Horse: 86%; Human: 100%.
Involved in sodium-dependent transport of D-mannose, D-glucose and D-fructose.
Specifications
SLC5A9 | |
Polyclonal | |
Unconjugated | |
SLC5A9 | |
hSGLT4; Na(+)/glucose cotransporter 4; SGLT4; SLC5A9; Sodium/glucose cotransporter 4; solute carrier family 5 (sodium/glucose cotransporter), member 9; solute carrier family 5 (sodium/sugar cotransporter), member 9; solute carrier family 5 member 9 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
200010 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q2M3M2 | |
SLC5A9 | |
synthetic peptide directed towards the N terminal region of human SLC5A9. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction