Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC6A12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557099
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557099 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557099 Supplier Invitrogen™ Supplier No. PA557099
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TRGPFRKRLR QLITPDSSLP QPKQHPCLDG SAGRNFGPSP TREGLIAGEK ETHL Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 57%.

Transports betaine and GABA. May have a role in regulation of GABAergic transmission in the brain through the reuptake of GABA into presynaptic terminals, as well as in osmotic regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC6A12
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC6A12
Gene Accession No. P48065
Gene Alias Betaine/GABA transporter 1; betaine/GABA transporter-1; BGT1; BGT-1; GABA transporter 1; Gabt2; gamma-aminobutyric acid (GABA-A) transporter 2; gamma-aminobutyric acid transporter; Gat1; GAT2; GAT-2; Na(+)/Cl(-) betaine/GABA transporter; RNU28927; SLC6A12; Sodium- and chloride-dependent betaine transporter; sodium- and chloride-dependent GABA transporter 2; solute carrier family 6 (neurotransmitter transporter), member 12; solute carrier family 6 (neurotransmitter transporter, betaine/GABA), member 12; solute carrier family 6 member 12; vesicular GABA transporter; VGAT
Gene Symbols SLC6A12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC6A12. Recombinant protein control fragment (Product #RP-95787).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6539
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.