Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC6A12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598160
Description
SLC6A12 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Transports betaine and GABA. May have a role in regulation of GABAergic transmission in the brain through the reuptake of GABA into presynaptic terminals, as well as in osmotic regulation.
Specifications
SLC6A12 | |
Polyclonal | |
Unconjugated | |
SLC6A12 | |
Betaine/GABA transporter 1; betaine/GABA transporter-1; BGT1; BGT-1; GABA transporter 1; Gabt2; gamma-aminobutyric acid (GABA-A) transporter 2; gamma-aminobutyric acid transporter; Gat1; GAT2; GAT-2; Na(+)/Cl(-) betaine/GABA transporter; RNU28927; SLC6A12; Sodium- and chloride-dependent betaine transporter; sodium- and chloride-dependent GABA transporter 2; solute carrier family 6 (neurotransmitter transporter), member 12; solute carrier family 6 (neurotransmitter transporter, betaine/GABA), member 12; solute carrier family 6 member 12; vesicular GABA transporter; VGAT | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6539 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.82 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P48065 | |
SLC6A12 | |
Recombinant Human Sodium- and chloride-dependent betaine transporter protein (515-614AA). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction