Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC6A12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142990
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142990 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142990 Supplier Invitrogen™ Supplier No. PA5142990
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.

Transports betaine and GABA. May have a role in regulation of GABAergic transmission in the brain through the reuptake of GABA into presynaptic terminals, as well as in osmotic regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC6A12
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene SLC6A12
Gene Accession No. P48065
Gene Alias Betaine/GABA transporter 1; betaine/GABA transporter-1; BGT1; BGT-1; GABA transporter 1; Gabt2; gamma-aminobutyric acid (GABA-A) transporter 2; gamma-aminobutyric acid transporter; Gat1; GAT2; GAT-2; Na(+)/Cl(-) betaine/GABA transporter; RNU28927; SLC6A12; Sodium- and chloride-dependent betaine transporter; sodium- and chloride-dependent GABA transporter 2; solute carrier family 6 (neurotransmitter transporter), member 12; solute carrier family 6 (neurotransmitter transporter, betaine/GABA), member 12; solute carrier family 6 member 12; vesicular GABA transporter; VGAT
Gene Symbols SLC6A12
Host Species Goat
Immunogen Peptide with sequence CLDGSAGRNFGPSPT.
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6539
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.