Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC6A12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA577453
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
Transports betaine and GABA. May have a role in regulation of GABAergic transmission in the brain through the reuptake of GABA into presynaptic terminals, as well as in osmotic regulation.
Specifications
SLC6A12 | |
Polyclonal | |
Unconjugated | |
Slc6a12 | |
betaine/GABA Transporter 1 (BGT-1) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14411, 50676 | |
-20°C | |
Lyophilized |
Western Blot | |
0.8 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
P31651, P48056 | |
Slc6a12 | |
Peptide (C)GEMMDQKDKDQVKD, corresponding to amino acid residues 22-35 of rat Betaine/GABA Transporter 1 | |
50 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction