Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC6A13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595662
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595662 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595662 Supplier Invitrogen™ Supplier No. PA595662
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC6A13 Polyclonal Antibody for Western Blot, IHC

Slc6a13 catalyzes sodium and chloride dependent transport of gamma-aminobutyric acid (GABA) to terminate GABA mediated synaptic activity. Slc6a13 is also a high-affinity gamma-aminobutyric acid (GABA) transporter, terminates biological actions of GABA by reuptake into cell, and its reuptake function utilizes sodium and chloride concentration gradients. Further, Slc6a13 may also aid in beta-alanine transport. Diseases associated with SLC6A13 include Traumatic Glaucoma and Neovascular Glaucoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC6A13
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation 10mM HEPES with 100μg/mL BSA, 50% glycerol, 150mM NaCl and no preservative; pH 7.5
Gene Slc6a13
Gene Accession No. P31646, P31649
Gene Alias GABA transport protein; GABA transporter 2; Gabt2; Gabt3; Gabt4; gamma-aminobutyric acid (GABA-A) transporter 3; GAT2; GAT-2; GAT3; Gat-3; Slc6a13; Sodium- and chloride-dependent GABA transporter 2; sodium- and chloride-dependent GABA transporter 3; solute carrier family 6 (neurotransmitter transporter), member 13; solute carrier family 6 (neurotransmitter transporter, GABA), member 13; solute carrier family 6 member 13
Gene Symbols Slc6a13
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acid residues from the C-terminal region conjugated to KLH.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14412, 171163
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.