Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC6A14 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP159659
Description
SLC6A14 Polyclonal specifically detects SLC6A14 in Human samples. It is validated for Western Blot.Specifications
SLC6A14 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
Amino acid transporter ATB0+, amino acid transporter B0+, BMIQ11, OBX, sodium- and chloride-dependent neutral and basic amino acid transporter B(0+), solute carrier family 6 (amino acid transporter), member 14, solute carrier family 6 (neurotransmitter transporter), member 14, Solute carrier family 6 member 14 | |
Rabbit | |
72 kDa | |
100 μL | |
Primary | |
Expected to cross react based on sequence identity: Guinea Pig 86%. | |
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q9UN76 | |
SLC6A14 | |
Synthetic peptides corresponding to SLC6A14(solute carrier family 6 (amino acid transporter), member 14) The peptide sequence was selected from the middle region of SLC6A14. Peptide sequence VFAGFAIFSILGHMAHISGKEVSQVVKSGFDLAFIAYPEALAQLPGGPFW The peptide sequence for this immunogen was taken from within the described region. | |
Affinity purified | |
RUO | |
11254 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction